Learn More
Agilent Technologies GLYKO SIALIDASE ASAMPLER KIT
Catalog No. NC0068520
Encompass
Try our lower MW isoforms for challenging sialidase digests.This Sampler Kit contains 1 vial each of 3 isoforms (88, 66 and 51 kDa) of the enzyme Sialidase A. Lower MW sialidases may help desialylate glycans if steric hindrance is an issue with regular sialidase.
Catalog No. NC0068520 Supplier Agilent Technologies Supplier No. GK80043
Try our lower MW isoforms for challenging sialidase digests.This Sampler Kit contains 1 vial each of 3 isoforms (88, 66 and 51 kDa) of the enzyme Sialidase A. Lower MW sialidases may help desialylate glycans if steric hindrance is an issue with regular sialidase.