Learn More
Agilent Technologies GLYKOCLEAN S CARTRIDGE
Catalog No. NC9080266
Encompass
Cartridgeused for clean up of glycan samples after fluorescent labeling. Also in combination with GlycoClean R to purify glycans from N-glycanase digests prior to Signal 2-AA or 2-AB labeling.
Catalog No. NC9080266 Supplier Agilent Technologies Supplier No. GKI4726
Cartridgeused for clean up of glycan samples after fluorescent labeling. Also in combination with GlycoClean R to purify glycans from N-glycanase digests prior to Signal 2-AA or 2-AB labeling.