Learn More
Agilent Technologies GLYKOCLEAN S-PLUS CARTRIDGES
Catalog No. NC1461909
Encompass
GlykoClean S-plus Cartridges [GC210]For clean-up of fluorescently labeled glycans. Mini cartridge comparable to our popular GlycoClean S Cartridge (product code GKI-4726) for use on the GlykoScreen Glycan Clean-up Station (product code GC100) vacuum manifold, 96-well array format.
Catalog No. NC1461909 Supplier Agilent Technologies Supplier No. GC210
GlykoClean S-plus Cartridges [GC210]For clean-up of fluorescently labeled glycans. Mini cartridge comparable to our popular GlycoClean S Cartridge (product code GKI-4726) for use on the GlykoScreen Glycan Clean-up Station (product code GC100) vacuum manifold, 96-well array format.