Learn More
Medchemexpress LLC Glyoxal | 107-22-2 | MFCD00006957 | 58.04 g/mol | C2H2O2 | 100mL
SDP
Catalog No. 5000269670
Encompass_Preferred
Glyoxal is a cytotoxic -oxoaldehyde Glyoxal induces cell damage and promotes protein glycation to form advanced glycation end-products (AGEs) Glyoxal is promising for research of oxidative stress-related diseases (such as atherosclerosis cataract Alzheimer s disease) and the formation of calcium oxalate kidney stones[1][2]
Catalog No. 50-002-69670 Supplier Medchemexpress LLC Supplier No. HYY0785100ML
Glyoxal is a cytotoxic -oxoaldehyde Glyoxal induces cell damage and promotes protein glycation to form advanced glycation end-products (AGEs) Glyoxal is promising for research of oxidative stress-related diseases (such as atherosclerosis cataract Alzheimer s disease) and the formation of calcium oxalate kidney stones[1][2]