Learn More
TRIBIOSCIENCE INC 40% Deionized Glyoxal Solution, 0.4ML
SDP
Supplier:  TRIBIOSCIENCE INC TBS6023
Encompass
40% Deionized Glyoxal Solution (6M) is designed for RNA denaturation compatible with all buffered agarose gels.
Catalog No. NC1795503