Learn More
Sigma Aldrich Fine Chemicals Biosciences Glyoxalase I Activity Assa
Catalog No. 502904145
Encompass
Glyoxalase I (GLO-1) a lactoylglutathione lyase also known as methylglyoxalase aldoketomutase ketone-aldehyde mutase and (R)-S-lactoylglutathione methylglyoxal-lyase is an enzyme that catalyzes the isomerization of hemithioacetal adducts which are formed in spontaneous reactions between glutathionyl groups and aldehydes. The primary physiological function of glyoxalase I is the detoxification of methyglyoxal a reactive 2-oxoaldehyde that is cytostatic at low concentrations and cytotoxic at millimolar concentrations. Glyoxalase I is a target for the development of pharmaceuticals against bacteria protozoans and cancer.
Catalog No. 50-290-4145 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK1141KT
Glyoxalase I (GLO-1) a lactoylglutathione lyase also known as methylglyoxalase aldoketomutase ketone-aldehyde mutase and (R)-S-lactoylglutathione methylglyoxal-lyase is an enzyme that catalyzes the isomerization of hemithioacetal adducts which are formed in spontaneous reactions between glutathionyl groups and aldehydes. The primary physiological function of glyoxalase I is the detoxification of methyglyoxal a reactive 2-oxoaldehyde that is cytostatic at low concentrations and cytotoxic at millimolar concentrations. Glyoxalase I is a target for the development of pharmaceuticals against bacteria protozoans and cancer.