Learn More
Medchemexpress LLC Glyoxalase I (lactoylglutathione lyase) | 9033-12-9 | >95.0% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP75162500UG
Encompass_Preferred
Recombinant mouse Glyoxalase I (GLO1) is supplied as a lyophilized, N-terminal 6xHis-tagged enzyme expressed in Escherichia coli for use in biochemical and enzymatic assays. The preparation is >95% pure, has a reported specific activity of 100.39 nmol/min/μg, and low endotoxin (<1 EU/μg). Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4; store at -20°C and aliquot after reconstitution.
Catalog No. 50-002-93035