Learn More
Medchemexpress LLC Glyoxalic acid | 298-12-4 | 50% | C2H2O3 | 500 G
SDP
Catalog No. 5000433176
Encompass_Preferred
Glyoxalic acid is an organic compound functioning as both an aldehyde and a carboxylic acid. It is supplied as a 50% w/w solution in water, appearing as a colorless to light yellow liquid. This compound is an endogenous metabolite and also identified as a human gut microbiota metabolite, intended for research use only.
  • Functions as both an aldehyde and a carboxylic acid
  • Induces fluorescence
  • Utilized in the study of neurons
  • Classified under ketones, aldehydes, and acids
  • Human endogenous metabolite
Catalog No. 50-004-33176 Supplier Medchemexpress LLC Supplier No. HY79494500G
Add to Cart
Add to Cart
Glyoxalic acid is an organic compound functioning as both an aldehyde and a carboxylic acid. It is supplied as a 50% w/w solution in water, appearing as a colorless to light yellow liquid. This compound is an endogenous metabolite and also identified as a human gut microbiota metabolite, intended for research use only.
  • Functions as both an aldehyde and a carboxylic acid
  • Induces fluorescence
  • Utilized in the study of neurons
  • Classified under ketones, aldehydes, and acids
  • Human endogenous metabolite