Learn More
Apexbio Technology LLC Glyoxylic acid 298-12-4 500mg
SDP
Catalog No. 5000064184
Encompass_Preferred
Glyoxylic acid (CAS 298-12-4) is a small organic molecule characterized structurally as both an aldehyde and a carboxylic acid Known as NSC 27785 in chemical databases glyoxylic acid participates biologically in various metabolic pathways notably serving as an intermediate in glyoxylate metabolism and glycine biosynthesis In biomedical research glyoxylic acid is extensively used as a reagent in chemical syntheses and employed experimentally to study metabolic fluxes and enzyme mechanisms related to energy metabolism and amino acid processing
Catalog No. 50-000-64184 Supplier Apexbio Technology LLC Supplier No. B7888500
Add to Cart
Add to Cart
Glyoxylic acid (CAS 298-12-4) is a small organic molecule characterized structurally as both an aldehyde and a carboxylic acid Known as NSC 27785 in chemical databases glyoxylic acid participates biologically in various metabolic pathways notably serving as an intermediate in glyoxylate metabolism and glycine biosynthesis In biomedical research glyoxylic acid is extensively used as a reagent in chemical syntheses and employed experimentally to study metabolic fluxes and enzyme mechanisms related to energy metabolism and amino acid processing