Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glyoxylic acid solution

Catalog No. 2601501KGA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-015-01KGA 1 kg
26-015-0250GA 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-015-01KGA Supplier Sigma Aldrich Supplier No. 2601501KGA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Glyoxylic acid has been employed:
  • As reducing agent in electroless copper depositions by free-formaldehyde method
  • In synthesis of new chelating agent, 2-(2-((2-hydroxybenzyl)amino)ethylamino)-2-(2-hydroxyphenyl)acetic acid (dcha)

Specifications

CAS 298-12-4
Density 1.342 g/mL (at 25°C)
Molecular Formula C2H2O3
Refractive Index n20/D 1.4149
Linear Formula OHCCOOH
MDL Number MFCD00006958
Quantity 1 kg
Synonym Formylformic acid; Oxoethanoic acid
Molecular Weight (g/mol) 74.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.