Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glyphosate-13C2,15N | 1185107-63-4 | 99.9% | 5 MG
SDP

Catalog No. 5000408164
Encompass_Preferred

Glyphosate-13C2,15N is the 13C and 15N labeled Glyphosate (HY-B0863). Glyphosate is an herbicidal derivative of the amino acid glycine. It targets and blocks the shikimate pathway in plants, a metabolic pathway not found in animals, which is essential for the synthesis of aromatic amino acids in plants.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Molecular weight: 172.05
  • Formula: C13C2H815NO5P
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: 13.89 mg/mL in H2O (80.73 mM; requires ultrasonic and warming)

Catalog No. 50-004-08164 Supplier Medchemexpress LLC Supplier No. HYB0863S35MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glyphosate-13C2,15N is the 13C and 15N labeled Glyphosate (HY-B0863). Glyphosate is an herbicidal derivative of the amino acid glycine. It targets and blocks the shikimate pathway in plants, a metabolic pathway not found in animals, which is essential for the synthesis of aromatic amino acids in plants.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Molecular weight: 172.05
  • Formula: C13C2H815NO5P
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: 13.89 mg/mL in H2O (80.73 mM; requires ultrasonic and warming)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.