Learn More
Chem Service Inc Glyphosate,1071-83-6,250MG
SDP
Catalog No. NC1029019
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula C3H8NO5P; Purity 98.70%; Molecular Weight 169.07; Melting Point 230 C; Synonyms: N-(Phosphonomethyl)glycine
Catalog No. NC1029019 Supplier Chem Service Inc Supplier No. N12133250MG
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula C3H8NO5P; Purity 98.70%; Molecular Weight 169.07; Melting Point 230 C; Synonyms: N-(Phosphonomethyl)glycine