Learn More
Medchemexpress LLC Glyphosate-d2-1 | 1071-83-6 | MFCD00055350 | 99.9% | 171.09 | C3H6D2NO5P | 10 MG
SDP
Catalog No. 5000298198
Encompass_Preferred
Glyphosate-d2-1 is a deuterium-labeled analytical standard of glyphosate supplied in milligram quantities for use in analytical and research applications. It is intended as an isotopically labeled internal standard to improve quantitation and identification in mass spectrometry and other analytical methods.
  • Deuterium-labeled glyphosate for use as an internal standard.
  • Supplied in milligram-scale quantity suitable for analytical workflows.
  • High listed purity for trace-level quantitation.
  • Accompanied by datasheet, certificate of analysis, and spectral reports.
  • Suitable for LC-MS/MS method development and validation.
Catalog No. 50-002-98198 Supplier Medchemexpress LLC Supplier No. HYB0863S210MG
Glyphosate-d2-1 is a deuterium-labeled analytical standard of glyphosate supplied in milligram quantities for use in analytical and research applications. It is intended as an isotopically labeled internal standard to improve quantitation and identification in mass spectrometry and other analytical methods.
  • Deuterium-labeled glyphosate for use as an internal standard.
  • Supplied in milligram-scale quantity suitable for analytical workflows.
  • High listed purity for trace-level quantitation.
  • Accompanied by datasheet, certificate of analysis, and spectral reports.
  • Suitable for LC-MS/MS method development and validation.