Learn More
Medchemexpress LLC Glyphosate-d2-1 | 99.9% | 5 MG
SDP
Catalog No. 5000409415
Encompass_Preferred
Glyphosate-d2-1 is a deuterium-labeled version of Glyphosate, an herbicidal derivative of the amino acid glycine. It targets and blocks the shikimate pathway in plants, a metabolic pathway not found in animals, which is essential for the synthesis of aromatic amino acids. This compound is primarily used as a tracer and internal standard in quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-09415 Supplier Medchemexpress LLC Supplier No. HYB0863S25MG
Glyphosate-d2-1 is a deuterium-labeled version of Glyphosate, an herbicidal derivative of the amino acid glycine. It targets and blocks the shikimate pathway in plants, a metabolic pathway not found in animals, which is essential for the synthesis of aromatic amino acids. This compound is primarily used as a tracer and internal standard in quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs