Learn More
Crescent Chemical Co Inc GLYPHOSATE-FMOC 50 MG
SDP
Supplier:  Crescent Chemical Co Inc PSCA14051000
Encompass_Preferred
Glyphosate-FMOC Herbicide (Metabolite neat) CAS 1373205-41-4 Molecular Weight - 391.3118 Storage - -18°C Catalog Num - DRE-CA14051000 pesticide
Catalog No. 50-777-19