Learn More
Medchemexpress LLC Glyphosate (Standard) | 1071-83-6 | 99.6% | 100 MG
SDP
Catalog No. 5000408861
Encompass_Preferred
Glyphosate (Standard) is the analytical standard of Glyphosate, an herbicidal derivative of the amino acid glycine. It targets and blocks the shikimate pathway, a plant metabolic pathway not found in animals, which is required for the synthesis of aromatic amino acids in plants. This product is intended for research and analytical applications.
  • Analytical standard for research
  • Reference standard for assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments including HPLC, GC, and MS
Catalog No. 50-004-08861 Supplier Medchemexpress LLC Supplier No. HYB0863R100MG
Glyphosate (Standard) is the analytical standard of Glyphosate, an herbicidal derivative of the amino acid glycine. It targets and blocks the shikimate pathway, a plant metabolic pathway not found in animals, which is required for the synthesis of aromatic amino acids in plants. This product is intended for research and analytical applications.
  • Analytical standard for research
  • Reference standard for assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments including HPLC, GC, and MS