Learn More
Medchemexpress LLC Glyphosate (Standard) | 1071-83-6 | 99.6% | 25 MG
SDP
Catalog No. 5000450651
Encompass_Preferred
Glyphosate (Standard) is an analytical standard intended for research and analytical applications. This herbicidal derivative of the amino acid glycine functions by targeting and blocking the shikimate pathway in plants, a metabolic process vital for the synthesis of aromatic amino acids that is not found in animals.
  • Analytical standard for research and analytical applications
  • Targets and blocks the shikimate pathway in plants
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments
  • Used in techniques such as HPLC, GC, and MS
Catalog No. 50-004-50651 Supplier Medchemexpress LLC Supplier No. 25MGHYB0863R
Glyphosate (Standard) is an analytical standard intended for research and analytical applications. This herbicidal derivative of the amino acid glycine functions by targeting and blocking the shikimate pathway in plants, a metabolic process vital for the synthesis of aromatic amino acids that is not found in animals.
  • Analytical standard for research and analytical applications
  • Targets and blocks the shikimate pathway in plants
  • Suitable for qualitative research experiments
  • Suitable for quantitative research experiments
  • Suitable for methodological research experiments
  • Used in techniques such as HPLC, GC, and MS