Learn More
Medchemexpress LLC Glyphosate (Standard) | 1071-83-6 | 99.6% | 250 MG
SDP
Catalog No. 5000406826
Encompass_Preferred
Glyphosate (Standard) is an analytical standard of Glyphosate, intended for research and analytical applications. This herbicidal derivative of the amino acid glycine specifically targets and blocks the plant shikimate pathway, crucial for aromatic amino acid synthesis. It is commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Analytical standard
  • Intended for research and analytical applications
  • Herbicidal derivative of amino acid glycine
  • Targets the shikimate pathway in plants
  • Commonly used in qualitative, quantitative, and methodological research
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-06826 Supplier Medchemexpress LLC Supplier No. 250MGHYB0863R
Glyphosate (Standard) is an analytical standard of Glyphosate, intended for research and analytical applications. This herbicidal derivative of the amino acid glycine specifically targets and blocks the plant shikimate pathway, crucial for aromatic amino acid synthesis. It is commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Analytical standard
  • Intended for research and analytical applications
  • Herbicidal derivative of amino acid glycine
  • Targets the shikimate pathway in plants
  • Commonly used in qualitative, quantitative, and methodological research
  • Suitable for HPLC, GC, and MS applications