Learn More
Medchemexpress LLC Glyphosate (Standard) | 1071-83-6 | 99.6% | 50 MG
SDP
Catalog No. 5000408903
Encompass_Preferred
Glyphosate (Standard) is an analytical reference standard. It is an herbicidal derivative of the amino acid glycine that targets and blocks the shikimate pathway in plants. This metabolic pathway, not found in animals, is necessary for the synthesis of aromatic amino acids in plants.
  • Used as a reference standard
  • Utilized in qualitative research experiments
  • Utilized in quantitative research experiments
  • Utilized in methodological research experiments
  • Commonly used in HPLC
  • Commonly used in GC
  • Commonly used in MS
Catalog No. 50-004-08903 Supplier Medchemexpress LLC Supplier No. HYB0863R50MG
Glyphosate (Standard) is an analytical reference standard. It is an herbicidal derivative of the amino acid glycine that targets and blocks the shikimate pathway in plants. This metabolic pathway, not found in animals, is necessary for the synthesis of aromatic amino acids in plants.
  • Used as a reference standard
  • Utilized in qualitative research experiments
  • Utilized in quantitative research experiments
  • Utilized in methodological research experiments
  • Commonly used in HPLC
  • Commonly used in GC
  • Commonly used in MS