Learn More
Pickering Laboratories Inc GLYPHOSPATE ANALYTICAL COLUMN
SDP
Supplier:  Pickering Laboratories Inc 1954150
Encompass_Preferred
Cation-exchange Column for Glyphosate analysis, 4 x 150 mm, including Glyphosate Test Mixture 1700-0080
Catalog No. NC9435095