Learn More
Biocare Medical, LLC GLYPICAN-3 6ML
SDP
Supplier:  Biocare Medical, LLC PM396AA
Encompass_Preferred
Studies show that Glypican-3 is expressed in most hepatocellular carcinomas (HCC), but is undetectable in normal liver and benign hepatic lesions, including dysplastic and cirrhotic nodules. It is a sensitive diagnostic marker for HCC.
Catalog No. 50-831-56