Learn More
Sino Biological Recombinant Human Glypican 3/GPC3 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10088H08HB100
Encompass_Preferred
A DNA sequence encoding the Human GPC3 (P51654-1) (Met1-His559) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6091