Learn More
ACROBiosystems ACROBiosystems Human Glypican 1 / GPC1 Protein, Fc Tag
Catalog No. 501956794
Encompass_Preferred
Recombinant Protein;1MG;Human Glypican 1, Fc Tag (GP1-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession # P35052-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 82.5 kDa. The protein migrates as 90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 μg/mL (100 μL/well) can bind Human Glypican 1, Fc Tag (Cat. No. GP1-H5254) with a linear range of 1-39 ng/mL (QC tested).
Catalog No. 50-195-6794 Supplier ACROBiosystems Supplier No. GP1H52541MG
Recombinant Protein;1MG;Human Glypican 1, Fc Tag (GP1-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession # P35052-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 82.5 kDa. The protein migrates as 90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 μg/mL (100 μL/well) can bind Human Glypican 1, Fc Tag (Cat. No. GP1-H5254) with a linear range of 1-39 ng/mL (QC tested).