Learn More
ACROBiosystems ACROBiosystems Mouse Glypican 1 / GPC1 Protein, His Tag
Supplier:  ACROBiosystems GP1M52H7100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse Glypican 1, His Tag (GP1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 529 (Accession # Q9QZF2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.5 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 μg/mL (100 μL/well) can bind Mouse Glypican 1, His Tag (Cat. No. GP1-M52H7) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-195-6773