Learn More
ACROBiosystems ACROBiosystems Human Glypican 2 / GPC2 Protein, Fc Tag
Catalog No. 501956477
Encompass_Preferred
Recombinant Protein;1MG;Human Glypican 2, Fc Tag (GP2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 553 (Accession # Q8N158-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.5 kDa.
Catalog No. 50-195-6477 Supplier ACROBiosystems Supplier No. GP2H52551MG
Recombinant Protein;1MG;Human Glypican 2, Fc Tag (GP2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 553 (Accession # Q8N158-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 84.5 kDa.