Learn More
ACROBiosystems ACROBiosystems Rhesus macaque Glypican 2 / GPC2 Protein, His Tag
Supplier:  ACROBiosystems GP2R52H9100UG
Encompass_Preferred
Recombinant Protein;100UG;Rhesus macaque Glypican 2, His Tag (GP2-R52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 552 (Accession # XP_001103286).;This protein carries a polyhistidine tag at the C-terminus.This protein contains a furin-like convertase cleavage site, 355-RNRR-358, and will be processed into N and C-terminal fragment with calculated MW of 37.3 kDa and 22.6 kDa respectively. The protein migrates as 26-30 kDa and 33-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Recombinant Human Midkine Protein at 2 μg/mL (100 μL/well) can bind Rhesus macaque Glypican 2, His Tag (Cat. No. GP2-R52H9) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-195-6476