Learn More
ACROBiosystems ACROBiosystems Mouse Glypican 2 / GPC2 Protein, His Tag
Supplier:  ACROBiosystems GP2M52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse Glypican 2, His Tag (GP2-M52H5) is expressed from human 293 cells (HEK293). It contains AA His 22 - Ser 556 (Accession # Q8BKV1-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.6 kDa. The protein migrates as 60 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Recombinant Human Midkine Protein, CF at 2 μg/mL (100 μL/well) can bind Mouse Glypican 2, His Tag (Cat. No. GP2-M52H5) with a linear range of 2-13 ng/mL (QC tested).
Catalog No. 50-195-6781