Learn More
ACROBiosystems ACROBiosystems Cynomolgus Glypican 3 / GPC3 Protein, Fc Tag
Catalog No. 501957388
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1).;This protein carries a human IgG1 Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 49.4 kDa respectively. The protein migrates as 40 kDa, 60 kDa and 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 2 μg/mL (100 μL/well) can bind Cynomolgus Glypican 3, Fc Tag (Cat. No. GP3-C5256) with a linear range of 1-31 ng/mL (QC tested).;Immobilized Cynomolgus Glypican 3, Fc Tag (Cat. No. GP3-C5256) at 0.5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GPC3 Antibody, Human IgG1 with a linear range of 0.2-3 ng/mL (Routinely tested).
Catalog No. 50-195-7388 Supplier ACROBiosystems Supplier No. GP3C52561MG
Recombinant Protein;1MG;Cynomolgus Glypican 3, Fc Tag (GP3-C5256) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1).;This protein carries a human IgG1 Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 49.4 kDa respectively. The protein migrates as 40 kDa, 60 kDa and 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 2 μg/mL (100 μL/well) can bind Cynomolgus Glypican 3, Fc Tag (Cat. No. GP3-C5256) with a linear range of 1-31 ng/mL (QC tested).;Immobilized Cynomolgus Glypican 3, Fc Tag (Cat. No. GP3-C5256) at 0.5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GPC3 Antibody, Human IgG1 with a linear range of 0.2-3 ng/mL (Routinely tested).