Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Mouse Glypican 3 Protein, Fc Tag, 100 μg

Catalog No. 501957395
Encompass_Preferred

Glypican-3 (GPC3) belongs to the glypican family and is highly expressed in lung, liver, and kidney tissues. This protein inhibits the dipeptidyl peptidase activity of DPP4 and plays a key role in the modulation of IGF2 interactions with its receptor. The protein is expressed in human HEK293 cells and contains a human IgG1 Fc tag at the C-terminus.

  • Greater than 90% purity as determined by SDS-PAGE
  • Expressed from human HEK293 cells
  • Lyophilized from 0.22 μm filtered solution containing Tris, glycine, arginine, and NaCl at pH 7.5 with trehalose
  • Contains AA Gln 25 to Arg 357 and Ser 358 to Met 557
  • Calculated molecular weight of 38.0 kDa and 49.1 kDa
  • Endotoxin level less than 1.0 EU per μg

Catalog No. 50-195-7395 Supplier ACROBiosystems Supplier No. GP3M5254100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glypican-3 (GPC3) belongs to the glypican family and is highly expressed in lung, liver, and kidney tissues. This protein inhibits the dipeptidyl peptidase activity of DPP4 and plays a key role in the modulation of IGF2 interactions with its receptor. The protein is expressed in human HEK293 cells and contains a human IgG1 Fc tag at the C-terminus.

  • Greater than 90% purity as determined by SDS-PAGE
  • Expressed from human HEK293 cells
  • Lyophilized from 0.22 μm filtered solution containing Tris, glycine, arginine, and NaCl at pH 7.5 with trehalose
  • Contains AA Gln 25 to Arg 357 and Ser 358 to Met 557
  • Calculated molecular weight of 38.0 kDa and 49.1 kDa
  • Endotoxin level less than 1.0 EU per μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.