Learn More
ACROBiosystems ACROBiosystems Mouse Glypican 3 / GPC3 Protein, Fc Tag
Supplier:  ACROBiosystems GP3M52541MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse Glypican 3, Fc Tag (GP3-M5254) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Met 557 (Accession # Q8CFZ4-1).;This protein carries a human IgG1 Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 354-RQYR-357, and will be partially processed into N and C-terminal fragment with calculated MW of 38.0 kDa and 49.1 kDa respectively. The protein migrates as 40 kDa, 60 kDa and 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 2 μg/mL (100 μL/well) can bind Mouse Glypican 3, Fc Tag (Cat. No. GP3-M5254) with a linear range of 1-10 ng/mL (QC tested).
Catalog No. 50-195-7394