Recombinant Protein;1MG;Human Glypican 3, Fc Tag, low endotoxin (GP3-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).;This protein carries a human IgG1 Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 49.3 kDa respectively. The protein migrates as 40 kDa, 60 kDa and 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 2 μg/mL (100 μL/well)can bind Human Glypican 3, Fc Tag (Cat. No. GP3-H5258) with a linear range of 4-63 ng/mL (QC tested).;Immobilized Human Glypican 3, Fc Tag, low endotoxin (Cat. No. GP3-H5258) at 0.5 μg/mL (100 μL/well) can bind Anti-Human GPC3 Mab with a linear range of 0.8-6 ng/mL (Routinely tested).