Learn More
ACROBiosystems ACROBiosystems Mouse Glypican 3 / GPC3 Protein, His Tag
Supplier:  ACROBiosystems GP3M52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse Glypican 3, His Tag (GP3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Met 557 (Accession # Q8CFZ4-1).;This protein carries a polyhistidine tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 354-RQYR-357, and will be partially processed into N and C-terminal fragment with calculated MW of 38.0 kDa and 24.4 kDa respectively. The protein migrates as 40 kDa and 66-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 0.5 μg/mL (100 μL/well) can bind Mouse Glypican 3, His Tag (Cat. No. GP3-M52H3) with a linear range of 1-16 ng/mL (QC tested).
Catalog No. 50-195-7398