Learn More
ACROBiosystems ACROBiosystems Cynomolgus Glypican 3 / GPC3 Protein, His Tag
Catalog No. 501957393
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus Glypican 3, His Tag (GP3-C5225) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1).;This protein carries a polyhistidine tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 24.6 kDa respectively. The N and C‑terminal fragment as 42 kDa and 66-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Glypican 3, His Tag (Cat. No. GP3-C5225) at 0.5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GPC3 Antibody, Human IgG1 with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-195-7393 Supplier ACROBiosystems Supplier No. GP3C5225100UG
Recombinant Protein;100UG;Cynomolgus Glypican 3, His Tag (GP3-C5225) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1).;This protein carries a polyhistidine tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 24.6 kDa respectively. The N and C‑terminal fragment as 42 kDa and 66-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Glypican 3, His Tag (Cat. No. GP3-C5225) at 0.5 μg/mL (100 μL/well) can bind Monoclonal Anti-Human GPC3 Antibody, Human IgG1 with a linear range of 0.2-3 ng/mL (QC tested).