Learn More
ACROBiosystems ACROBiosystems Cynomolgus Glypican 3 / GPC3 Protein, Llama IgG2b Fc Tag, low endotoxin
Catalog No. 501956767
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (GP3-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1).;This protein carries a llama IgG2b Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 50.5 kDa respectively. The protein migrates as 40 kDa, 60 kDa and 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 μg/mL (100 μL/well) can bind Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. GP3-C5259) with a linear range of 1-63 ng/mL (QC tested).
Catalog No. 50-195-6767 Supplier ACROBiosystems Supplier No. GP3C5259100UG
Recombinant Protein;100UG;Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (GP3-C5259) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # XP_005594665.1).;This protein carries a llama IgG2b Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa and 50.5 kDa respectively. The protein migrates as 40 kDa, 60 kDa and 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 μg/mL (100 μL/well) can bind Cynomolgus Glypican 3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. GP3-C5259) with a linear range of 1-63 ng/mL (QC tested).