Learn More
ACROBiosystems Recombinant Protein;FITC-Labeled Human Glypican 3 / GPC3 Protein, Fc Tag;HEK293;25UG;GP3,GPC3,OCI5,Glypican-3,GPC-3,GTR2-2,Glypican 3,MXR7,DGSX ,MXR-7,OCI-5 ,SDYS ,SGB,SGBS,SGBS1
Catalog No. 501657585
Encompass_Preferred
Recombinant Protein;FITC-Labeled Human Glypican 3 / GPC3 Protein, Fc Tag;25UG;FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (Accession # P51654). It is the FITC labeled form of Human Glypican 3, Fc Tag (Cat. No. GP3-H5258).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 87.4 kDa. The protein migrates as 43 kDa, 58 kDa and 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human GPC3 MAb at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) with a linear range of 0.078-1.25 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).;293 cells were transfected with anti-GPC3-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human Glypican 3 Protein, Fc Tag (Cat. No. GP3-HF258, 3 µg/ml) and C. FITC-labeled Protein Control. A. Non-transfected 293 cells and C. FITC-labeled
Catalog No. 50-165-7585 Supplier ACROBiosystems Supplier No. GP3HF25825UG
Recombinant Protein;FITC-Labeled Human Glypican 3 / GPC3 Protein, Fc Tag;25UG;FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) is expressed from human HEK293 cells. It contains AA Gln 25 - His 559 (Accession # P51654). It is the FITC labeled form of Human Glypican 3, Fc Tag (Cat. No. GP3-H5258).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 87.4 kDa. The protein migrates as 43 kDa, 58 kDa and 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human GPC3 MAb at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human Glypican 3, Fc Tag (Cat. No. GP3-HF258) with a linear range of 0.078-1.25 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).;293 cells were transfected with anti-GPC3-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human Glypican 3 Protein, Fc Tag (Cat. No. GP3-HF258, 3 µg/ml) and C. FITC-labeled Protein Control. A. Non-transfected 293 cells and C. FITC-labeled