Learn More
ACROBiosystems Recombinant Protein;Human Glypican 3 / GPC3 Protein;HEK293;1MG;GP3,GPC3,OCI5,Glypican-3,GPC-3,GTR2-2,Glypican 3,MXR7,DGSX ,MXR-7,OCI-5 ,SDYS ,SGB,SGBS,SGBS1
Catalog No. 501657465
Encompass_Preferred
Recombinant Protein;Human Glypican 3 / GPC3 Protein;1MG;Human Glypican 3 (GP3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 62.7 kDa. The protein migrates as 30-33 kDa, 40-44 kDa and 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 0.5 μg/mL (100 μL/well) can bind Human Glypican 3 (Cat. No. GP3-H52H4) with a linear range of 8-31 ng/mL (QC tested).;Immobilized Human Glypican 3 (Cat. No. GP3-H52H4) at 0.5 μg/mL (100 μL/well) can bind Anti-Human GPC3 Mab with a linear range of 0.1-1.6 ng/mL (Routinely tested).
Catalog No. 50-165-7465 Supplier ACROBiosystems Supplier No. GP3H52H41MG
Recombinant Protein;Human Glypican 3 / GPC3 Protein;1MG;Human Glypican 3 (GP3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 62.7 kDa. The protein migrates as 30-33 kDa, 40-44 kDa and 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 0.5 μg/mL (100 μL/well) can bind Human Glypican 3 (Cat. No. GP3-H52H4) with a linear range of 8-31 ng/mL (QC tested).;Immobilized Human Glypican 3 (Cat. No. GP3-H52H4) at 0.5 μg/mL (100 μL/well) can bind Anti-Human GPC3 Mab with a linear range of 0.1-1.6 ng/mL (Routinely tested).