Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Glypican 3 / GPC3 Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;GP3,GPC3,OCI5,Glypican-3,GPC-3,GTR2-2,Glypican 3,MXR7,DGSX ,MXR-7,OCI-5 ,SDYS ,SGB,SGBS,SGBS1
Supplier:  ACROBiosystems GP3H82E5200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human Glypican 3 / GPC3 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human Glypican 3, His Tag, Avi Tag (GP3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 64.5 kDa. The protein migrates as 42 kDa(caused by a furin cleavage site) and 70-250 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Glypican 3, His Tag, Avi Tag (Cat. No. GP3-H82E5) with a linear range of 1-16 ng/mL (QC tested).;Immobilized Anti-Human GPC3 Mab at 2 μg/mL (100 μL/well) can bind Biotinylated Human Glypican 3, His Tag, Avi Tag (Cat. No. GP3-H82E5) with a linear range of 0.4-6 ng/mL (Routinely tested).;293 cells were transfected with anti-GPC3-scFv and RFP ta
Catalog No. 50-165-8076