Learn More
Cayman Chemical GlypIcn3 RabbIt MoncLNal
SDP
Catalog No. 502919213
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to the central region of human glypican-3 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Glypican-3 • Applications: FC
Catalog No. 50-291-9213 Supplier Cayman Chemical Supplier No. 39540100 TEST
Immunogen: A synthetic peptide corresponding to the central region of human glypican-3 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Glypican-3 • Applications: FC