Learn More
Cayman Chemical Glypican-3 Rabbit Monoclonal Antibody (APC) (Clone 024), 25 test
SDP
Catalog No. 502919214
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to the central region of human glypican-3 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Glypican-3 • Applications: FC
Catalog No. 50-291-9214 Supplier Cayman Chemical Supplier No. 3954025 TEST
Immunogen: A synthetic peptide corresponding to the central region of human glypican-3 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Glypican-3 • Applications: FC