Learn More
Cayman Chemical GlypIcn3 RabbIt MoncLNal 25tSt
SDP
Supplier:  Cayman Chemical 3700825 TEST
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to the central region of human glypican-3 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Glypican-3 • Applications: FC
Catalog No. 50-291-5476