Learn More
Medchemexpress LLC HY-108940 10mg Medchemexpress, GlyRS-IN-1 CAS:112921-11-6 Purity:>98%
SDP
Catalog No. 501968636
Encompass_Preferred
Medchemexpress, HY-108940 10mg GlyRS-IN-1 CAS:112921-11-6 GlyRS-IN-1 is a glycyl-tRNA synthase (GlyRS) inhibitor extracted from patent WO 2017066459 A1. GlyRS-IN-1 can also inhibit the growth of bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8636 Supplier Medchemexpress LLC Supplier No. HY10894010MG
Medchemexpress, HY-108940 10mg GlyRS-IN-1 CAS:112921-11-6 GlyRS-IN-1 is a glycyl-tRNA synthase (GlyRS) inhibitor extracted from patent WO 2017066459 A1. GlyRS-IN-1 can also inhibit the growth of bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.