Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GlyT1 Inhibitor 1 | 1820934-93-7 | 98.59% | 387.43 | 10 MG
SDP

Catalog No. 5000438379
Encompass_Preferred

GlyT1 Inhibitor 1 is a potent and selective GlyT1 inhibitor with an IC50 of 38 nM for rGlyT1. It also demonstrates antipsychotic activity and in vivo efficacy in a rat novel object recognition (NOR) assay after oral dosing at 0.1, 1, and 3 mg/kg.

  • Potent and selective GlyT1 inhibitor
  • IC50: 38 nM for rGlyT1
  • Antipsychotic activity
  • In vivo efficacy in rat novel object recognition (NOR) assay
  • Appearance: Solid, White to off-white
  • Formula: C22H21N5O2
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; In solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility (In Vitro): DMSO : 25 mg/mL (64.53 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Catalog No. 50-004-38379 Supplier Medchemexpress LLC Supplier No. HY11243210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GlyT1 Inhibitor 1 is a potent and selective GlyT1 inhibitor with an IC50 of 38 nM for rGlyT1. It also demonstrates antipsychotic activity and in vivo efficacy in a rat novel object recognition (NOR) assay after oral dosing at 0.1, 1, and 3 mg/kg.

  • Potent and selective GlyT1 inhibitor
  • IC50: 38 nM for rGlyT1
  • Antipsychotic activity
  • In vivo efficacy in rat novel object recognition (NOR) assay
  • Appearance: Solid, White to off-white
  • Formula: C22H21N5O2
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; In solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility (In Vitro): DMSO : 25 mg/mL (64.53 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.