Learn More
Medchemexpress LLC GlyT1 Inhibitor 1 | 1820934-93-7 | 98.6% | 387.43 | 50 MG
SDP
Catalog No. 5000438521
Encompass_Preferred
GlyT1 Inhibitor 1 is a potent and selective inhibitor of GlyT1, with an IC50 of 38 nM for rGlyT1. It also demonstrates antipsychotic activity and in vivo efficacy in a rat novel object recognition (NOR) assay after oral dosing at 0.1, 1, and 3 mg/kg. This product is for research use only and not sold to patients.
  • Potent and selective GlyT1 inhibitor.
  • IC50 of 38 nM for rGlyT1.
  • Demonstrates antipsychotic activity.
  • Shows in vivo efficacy in a rat novel object recognition (NOR) assay.
Catalog No. 50-004-38521 Supplier Medchemexpress LLC Supplier No. HY11243250MG
GlyT1 Inhibitor 1 is a potent and selective inhibitor of GlyT1, with an IC50 of 38 nM for rGlyT1. It also demonstrates antipsychotic activity and in vivo efficacy in a rat novel object recognition (NOR) assay after oral dosing at 0.1, 1, and 3 mg/kg. This product is for research use only and not sold to patients.
  • Potent and selective GlyT1 inhibitor.
  • IC50 of 38 nM for rGlyT1.
  • Demonstrates antipsychotic activity.
  • Shows in vivo efficacy in a rat novel object recognition (NOR) assay.