Learn More
Apexbio Technology LLC GM 6001 142880-36-2 5mg
SDP
Catalog No. 5000064050
Encompass_Preferred
GM 6001 (Ilomastat Galardin CAS 142880-36-2) is a broad-spectrum inhibitor of matrix metalloproteinases (MMPs) including MMP-1 -2 -3 -8 and -9 with reported Ki values of 0 4 nM 0 5 nM 27 nM 0 1 nM and 0 2 nM respectively MMPs participate in extracellular matrix remodeling and mediate signaling events induced by GPCR agonists through EGFR transactivation GM 6001 suppresses EGFR phosphorylation ERK activation and subsequent DNA synthesis induced by stimuli such as bombesin and lysophosphatidic acid (LPA) Additionally this compound has potential applications in cartilage research facilitating meniscal repair under inflammatory conditions by dampening IL-1-mediated MMP activity
Catalog No. 50-000-64050 Supplier Apexbio Technology LLC Supplier No. A40505
GM 6001 (Ilomastat Galardin CAS 142880-36-2) is a broad-spectrum inhibitor of matrix metalloproteinases (MMPs) including MMP-1 -2 -3 -8 and -9 with reported Ki values of 0 4 nM 0 5 nM 27 nM 0 1 nM and 0 2 nM respectively MMPs participate in extracellular matrix remodeling and mediate signaling events induced by GPCR agonists through EGFR transactivation GM 6001 suppresses EGFR phosphorylation ERK activation and subsequent DNA synthesis induced by stimuli such as bombesin and lysophosphatidic acid (LPA) Additionally this compound has potential applications in cartilage research facilitating meniscal repair under inflammatory conditions by dampening IL-1-mediated MMP activity