Learn More
Abcam GM 6001, MMP inhibitor, 1MG
Catalog No. 502584558
Encompass_Preferred
MW 388.5 Da, Purity >98%. Broad spectrum potent MMP inhibitor. Ki values are 0.4 (MMP-1), 0.5 (MMP-2), 27 (MMP-3), 0.1 (MMP-8), 0.2 (MMP-9), 13.4 (MMP-14) and 0.36 (MMP-26) nM. IC₅₀ value for MMP-7 is 3.7 nM. Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4558 Supplier Abcam Supplier No. AB1208451MG
MW 388.5 Da, Purity >98%. Broad spectrum potent MMP inhibitor. Ki values are 0.4 (MMP-1), 0.5 (MMP-2), 27 (MMP-3), 0.1 (MMP-8), 0.2 (MMP-9), 13.4 (MMP-14) and 0.36 (MMP-26) nM. IC₅₀ value for MMP-7 is 3.7 nM. Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement