Learn More
Enzo Life Sciences GM-6001, (1mg), CAS Number: 142880-36-2
SDP
Catalog No. NC9924174
Encompass_Preferred
MMP inhibitor. Potent broad-spectrum hydroxamate inhibitor of matrix metalloproteinases (MMPs). .in vivo. Alternative Name: Galardin, Ilomastat, N-[(2R)-2-(Hydroxamidocarbonylmethyl)-4-methylpentanoyl]-L-tryptophan methylamide. Formula: C20H28N404. MW: 388.5. CAS Number: 142880-36-2. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (at least 25mM) or 100% ethanol (10mg/ml). Long Term Storage: -20°C
Catalog No. NC9924174 Supplier Enzo Life Sciences Supplier No. BMLEI3000001
MMP inhibitor. Potent broad-spectrum hydroxamate inhibitor of matrix metalloproteinases (MMPs). .in vivo. Alternative Name: Galardin, Ilomastat, N-[(2R)-2-(Hydroxamidocarbonylmethyl)-4-methylpentanoyl]-L-tryptophan methylamide. Formula: C20H28N404. MW: 388.5. CAS Number: 142880-36-2. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (at least 25mM) or 100% ethanol (10mg/ml). Long Term Storage: -20°C