Learn More
Medchemexpress LLC GM-60186 | 99.3% | 504.59 | 1 ML
SDP
Catalog No. 5000390951
Encompass_Preferred
GM-60186 is a 5-HT receptor 2B (HTR2B) inhibitor with an IC50 of 257 nM. It significantly inhibits colorectal cancer proliferation and migration.
  • Inhibits HTR2B with an IC50 of 257 nM.
  • Demonstrates anti-colorectal cancer activity.
  • In vitro, it effectively inhibits proliferation in colorectal cancer (CRC) cells (COLO-205, MC38, CT26, and Caco2 cells) at concentrations of 50-500 nM over 72 hours.
  • Effectively inhibits colorectal cancer migration at 500 nM over 24-48 hours.
  • In vivo, it significantly reduces tumor size and weight in xenograft mouse models at doses of 10-20 mg/kg, administered daily for 20 days.
Catalog No. 50-003-90951 Supplier Medchemexpress LLC Supplier No. HY16922910MM/1ML
GM-60186 is a 5-HT receptor 2B (HTR2B) inhibitor with an IC50 of 257 nM. It significantly inhibits colorectal cancer proliferation and migration.
  • Inhibits HTR2B with an IC50 of 257 nM.
  • Demonstrates anti-colorectal cancer activity.
  • In vitro, it effectively inhibits proliferation in colorectal cancer (CRC) cells (COLO-205, MC38, CT26, and Caco2 cells) at concentrations of 50-500 nM over 72 hours.
  • Effectively inhibits colorectal cancer migration at 500 nM over 24-48 hours.
  • In vivo, it significantly reduces tumor size and weight in xenograft mouse models at doses of 10-20 mg/kg, administered daily for 20 days.