Learn More
Medchemexpress LLC GM-CSF 2ug
SDP
Catalog No. 5000181096
Encompass_Preferred
GM-CSF R alpha is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2 GM-CSF R alpha binds to the cytokine with high specificity and low affinity[1] GM-CSF R alpha binds to GM-CSF and induces cell differentiation[2] GM-CSF R alpha monoclonal antibody decreases GM-CSFR expression on GM-CSF-responsive cells and shows anti-inflammatory activity in rheumatoid arthritis (RA)[3] GM-CSF R alpha Protein Human (HEK293 Fc) is a recombinant protein with a C-Terminal Fc label It consists of 320 amino acids (M1-G320) and is produced in HEK293 cells
Catalog No. 50-001-81096 Supplier Medchemexpress LLC Supplier No. HYP730832UG
GM-CSF R alpha is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2 GM-CSF R alpha binds to the cytokine with high specificity and low affinity[1] GM-CSF R alpha binds to GM-CSF and induces cell differentiation[2] GM-CSF R alpha monoclonal antibody decreases GM-CSFR expression on GM-CSF-responsive cells and shows anti-inflammatory activity in rheumatoid arthritis (RA)[3] GM-CSF R alpha Protein Human (HEK293 Fc) is a recombinant protein with a C-Terminal Fc label It consists of 320 amino acids (M1-G320) and is produced in HEK293 cells