Learn More
Medchemexpress LLC GM-CSF Canine 50ug | 50ug
SDP
Catalog No. 5000194586
Encompass_Preferred
The CSF2 protein is a cytokine that plays a critical role in the regulation of immune responses and hematopoiesis It stimulates the production and differentiation of white blood cells including macrophages and granulocytes GM-CSF Protein Canine is the recombinant canine-derived GM-CSF protein expressed by E coli with tag free
Catalog No. 50-001-94586 Supplier Medchemexpress LLC Supplier No. HYP7908750UG
The CSF2 protein is a cytokine that plays a critical role in the regulation of immune responses and hematopoiesis It stimulates the production and differentiation of white blood cells including macrophages and granulocytes GM-CSF Protein Canine is the recombinant canine-derived GM-CSF protein expressed by E coli with tag free