Learn More
Enzo Life Sciences GM-CSF (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2520100
Encompass_Preferred
Alternative name: Granulocyte-macrophage colony stimulating factor, CSF-2, Pluripoietin-alpha, MGI1GM. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium phosphate, pH 7.5. Source: Produced in E. coli. Contains 128 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4068